Pharmaceutical secondary standard, Tazobactam, certified reference material, Supelco®
PHR1686-1G
: Merck
:
- Concentration:Neat
- Pk:1 g
- Standards:Pharmaceutical secondary standard (CRM)
- CAS No.:89786-04-9
- Description:Tazobactam
- Description:Tazobactam
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.