Pharmaceutical secondary standard, Tartaric Acid, certified reference material, Supelco®
PHR1472-1G
: Merck
:
- Koncentration:Neat
- Förp.:1 g
- Standard:Pharmaceutical secondary standard (CRM)
- CAS-nr:87-69-4
- Beskrivning:Tartaric Acid
- Benämning:Tartaric Acid
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.