Order Entry
Sweden
ContactUsLinkComponent
 
Pharmaceutical secondary standard, Monobasic Potassium Phosphate, certified reference material, Supelco®
  PHR1330-5G
 :  Merck
 :  
undefined
Pharmaceutical secondary standard, Monobasic Potassium Phosphate, certified reference material, Supelco®
  PHR1330-5G
 :  Merck
 :  

 

  • Concentration:
    Neat
  • Pk:
    5 g
  • Standards:
    Pharmaceutical secondary standard (CRM)
  • CAS No.:
    7778-77-0
  • Description:
    Monobasic Potassium Phosphate
  • Description:
    Monobasic Potassium Phosphate

 

 

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

 : This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.