Order Entry
Sweden
ContactUsLinkComponent
Pharmaceutical secondary standard, Benzocaine, certified reference material, Supelco®
Pharmaceutical secondary standard, Benzocaine, certified reference material, Supelco®
Catalog # PHR1158-1G
Supplier:  Merck
CAS Number:  
undefined
Pharmaceutical secondary standard, Benzocaine, certified reference material, Supelco®
Catalog # PHR1158-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Pk:
    1 g
  • Standards:
    Pharmaceutical secondary standard (CRM)
  • CAS No.:
    94-09-7
  • Description:
    Benzocaine
  • Description:
    Benzocaine

Specifications

About this item

Traceable to USP 1054000.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.