Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Stirrers
Magnetic Stirring Bars
Magnetic stirring bar, WHEATON®
Magnetic stirring bar, WHEATON®
Magnetic stirring bar, WHEATON®
Katalog # WHEA903007
Leverantör:
DWK Life Sciences
Expand image
Magnetic stirring bar, WHEATON®
Katalog # WHEA903007
Leverantör:
DWK Life Sciences
Specifikationer
For:
Micro Kit®
Width:
3 mm
Length:
10 mm
Pk:
6
Specifikationer
Om denna produkt
Product Details
Product Details