Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Indapamide, for drug analysis, analytical standard, Supelco®
Indapamide, for drug analysis, analytical standard, Supelco®
Indapamide, for drug analysis, analytical standard, Supelco®
Supplier:
Merck
Certificates
Expand image
Indapamide, for drug analysis, analytical standard, Supelco®
Supplier:
Merck
About this item
Specifications
CAS No.:
26807-65-8
M-Clarity quality level:
MQ100
Description:
Indapamide
Specifications