Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Chlorpropamide, analytical standard, Supelco®
Chlorpropamide, analytical standard, Supelco®
Chlorpropamide, analytical standard, Supelco®
Catalog # C1290-25G
Leverantör:
Merck
CAS-nummer:
Expand image
Chlorpropamide, analytical standard, Supelco®
Catalog # C1290-25G
Leverantör:
Merck
CAS-nummer:
Specifikationer
Pk:
25 g
Standards:
analytical standard
CAS No.:
94-20-2
Description:
Chlorpropamide
Description:
Chlorpropamide
Specifikationer
Om denna produkt
Product Details
Product Details