0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Standards
Organic Standards
Oxibendazole, analytical standard, VETRANAL™, Supelco®
Oxibendazole, analytical standard, VETRANAL™, Supelco®
32924-100MG
:
Merck
:
Oxibendazole, analytical standard, VETRANAL™, Supelco®
32924-100MG
:
Merck
:
Pk:
100 mg
Standards:
analytical standard
CAS No.:
20559-55-1
Description:
Oxibendazole
M-Clarity quality level:
MQ100
Description:
Oxibendazole