Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standarder
Organic Standards
Ofloxacin-d3, analytical standard, VETRANAL™, Supelco®
Ofloxacin-d3, analytical standard, VETRANAL™, Supelco®
32436-10MG
:
Merck
:
Ofloxacin-d3, analytical standard, VETRANAL™, Supelco®
32436-10MG
:
Merck
:
Förp.:
10 mg
Standard:
analytical standard
CAS-nr:
1173147-91-5
Beskrivning:
Ofloxacin-d3
Benämning:
Ofloxacin-d3