0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Standards
Organic Standards
Sulfadoxin, analytical standard, VETRANAL™, Supelco®
Sulfadoxin, analytical standard, VETRANAL™, Supelco®
31736-250MG
:
Merck
:
Sulfadoxin, analytical standard, VETRANAL™, Supelco®
31736-250MG
:
Merck
:
Pk:
250 mg
Standards:
analytical standard
CAS No.:
2447-57-6
Description:
Sulfadoxin
M-Clarity quality level:
MQ100
Description:
Sulfadoxin