Order Entry
Sweden
ContactUsLinkComponent
 
Corrosive
Environment
Silver plating stain kit acc. to von Kossa for calcium determination, stain for histology, Sigma-Aldrich®
Catalog # 1.00362.0001
Supplier:  Merck
CAS Number:  
Corrosive
Environment
Silver plating stain kit acc. to von Kossa for calcium determination, stain for histology, Sigma-Aldrich®
Catalog # 1.00362.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Fibre case
  • Pk:
    1 SET
  • Seq:
    0125
  • Pk Info:
    Preparation Capacity 300 slides

Specifications

Specification Test Results

  • Suitability for microscopy:
    passes test
  • Calcium:
    brown to black
  • Nuclei:
    red
  • Collagen:
    red
  • Background:
    red

Specification Test Results

About this item

 :
Storage Temperature:Ambient
UN:3316
ADR:9,II
 :GHS05
 :GHS09
 :1