Order Entry
Sweden
ContactUsLinkComponent
Organic reference standard, Thiamine Hydrochloride (B1), analytical standard, Supelco®
Organic reference standard, Thiamine Hydrochloride (B1), analytical standard, Supelco®
Catalog # 47858.
Supplier:  Merck
CAS Number:  
Organic reference standard, Thiamine Hydrochloride (B1), analytical standard, Supelco®
Catalog # 47858.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • CAS No.:
    67-03-8
  • Description:
    Thiamine Hydrochloride (B1)
  • Melting point:
    250 °C
  • Formula:
    C₁₀H₁₆
  • Storage temperature:
    Ambient
  • Description:
    Thiamine Hydrochloride (B1)
  • Pk Info:
    Ampoule of 1000 mg
  • Form:
    Neat

Specifications

About this item

Thiamine hydrochloride (B1) is a water soluble vitamin, which belongs the B-complex group of vitamins. It can exist both as a hemihydrate and as a nonstoichiometric hydrate. It can generally act like a co-enzyme for biochemical reactions involved in carbohydrate metabolism.

It can exist both as a hemihydrate and as a nonstoichiometric hydrate. It can generally act like a co-enzyme for biochemical reactions involved in carbohydrate metabolism.