Supelco® Organic Reference Standard, Kaempferide
Catalog # 69545-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:491-54-3
- Description:Kaempferide
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD00016771
- M-Clarity quality level:MQ100
- Synonym(s):3,5,7-Trihydroxy-4′-methoxyflavone; KF; 4′-Methoxy-3,5,7-trihydroxyflavone
- Molecular weight:300,26