Supelco® Organic Reference Standard, Isorhamnetin
Catalog # 38907-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:480-19-3
- Description:Isorhamnetin
- Storage conditions:2...8 °C
- Shipping conditions:WICE
- MDL No.:MFCD00017310
- M-Clarity quality level:MQ100
- Synonym(s):3′-Methoxy-3,4′,5,7-tetrahydroxyflavone; 3′-Methylquercetin; 3,4′,5,7-Tetrahydroxy 3′-methoxyflavone; Isorhamnetol; Quercetin 3′-methyl ether
- Formula:C₁₆H₁₂O₇
- Molecular weight:316,26
- Storage temperature:2...8 °C
- Flash point:Not applicable
- Form:Neat
Specifications
About this item
- GHS signal: GHS08
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.