Order Entry
Sweden
ContactUsLinkComponent
 
Organic reference standard, Isoeugenol, predominantly trans, analytical standard, Supelco®
Catalog # 57522-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Isoeugenol, predominantly trans, analytical standard, Supelco®
Catalog # 57522-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    5932-68-3
  • Description:
    Isoeugenol, predominantly trans, ≥99,0%
  • Formula:
    C₁₀H₁₂O₂
  • Description:
    Isoeugenol

Specifications

About this item