Supelco® Organic Reference Standard, Hexahydrocurcumin
Catalog # 69727-10MG
Leverantör: Merck
CAS-nummer:
Specifikationer
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:36062-05-2
- Description:Hexahydrocurcumin
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD18783096
- M-Clarity quality level:MQ100
- Synonym(s):(RS)-5-Hydroxy-1,7-bis(4-hydroxy-3-methoxyphenyl)-3-heptanone
- Molecular weight:374,43
- Flash point:218 °C