Order Entry
Sweden
ContactUsLinkComponent
Supelco® Organic Reference Standard, Hesperidin
Supelco® Organic Reference Standard, Hesperidin
Catalog # 50162-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Hesperidin
Catalog # 50162-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    520-26-3
  • Description:
    Hesperidin
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00075663
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3′,5,7-Trihydroxy 4′-methoxyflavanone 7-rutinoside; Hesperetin 7-rhamnoglucoside; Hesperitin-7-rutinoside
  • Melting point:
    250 - 255 °C (dec.) (lit.)
  • Molecular weight:
    610,56

Specifications

About this item