Order Entry
Sweden
ContactUsLinkComponent
Supelco® Organic Reference Standard, Harpagoside
Supelco® Organic Reference Standard, Harpagoside
Catalog # 68527-10MG
Leverantör:  Merck
CAS-nummer:  
undefined
Supelco® Organic Reference Standard, Harpagoside
Catalog # 68527-10MG
Leverantör:  Merck
CAS-nummer:  

Specifikationer

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    19210-12-9
  • Description:
    Harpagoside
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00017415
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    494,49

Specifikationer

Om denna produkt

  • GHS signal: GHS07