Order Entry
Sweden
ContactUsLinkComponent
Supelco® Organic Reference Standard, γ-Carotene
Supelco® Organic Reference Standard, γ-Carotene
Catalog # 05387-1MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, γ-Carotene
Catalog # 05387-1MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Standards:
    Analytical standard
  • CAS No.:
    472-93-5
  • Description:
    γ-Carotene
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    536,87
  • Storage temperature:
    −20 °C
  • Flash point:
    Not applicable

Specifications

About this item