Supelco® Organic Reference Standard, Bisdemethoxycurcumin
Catalog # 90594-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:33171-05-0
- Description:Bisdemethoxycurcumin
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD03419284
- M-Clarity quality level:MQ100
- Synonym(s):(E,E)-1,7-Bis(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione; Bisdesmethoxycurcumin; Curcumin III; Didemethoxycurcumin
- Molecular weight:308,33