Supelco® Organic Reference Standard, Avenanthramide C
36465-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:116764-15-9
- Description:Avenanthramide C
- Storage conditions:2...8 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD23380831
- M-Clarity quality level:MQ100
- Synonym(s):2-{[(2E)-3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]amino}-5-hydroxybenzoic acid; N-[3′,4′-Dihydroxy-(E)-cinnamoyl]-5-hydroxyanthranilic acid; Avenanthramide Bc
- Formula:C₁₆H₁₃NO₆
- Molecular weight:315,28
- Storage temperature:2...8 °C
- Flash point:Not applicable
- Form:Neat
- GHS signal: GHS07