Order Entry
Sweden
ContactUsLinkComponent
Organic reference standard, (2RS)-2-(4-Hydroxyphenyl)propionic acid, ≥95,0% (qNMR), Supelco®
Organic reference standard, (2RS)-2-(4-Hydroxyphenyl)propionic acid, ≥95,0% (qNMR), Supelco®
  91378-10MG
 :  Merck
 :  
Organic reference standard, (2RS)-2-(4-Hydroxyphenyl)propionic acid, ≥95,0% (qNMR), Supelco®
  91378-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • CAS No.:
    938-96-5
  • Description:
    (2RS)-2-(4-Hydroxyphenyl)propionic acid,≥98,0% (HPLC)
  • Seq:
    01
  • Formula:
    C₉H₁₀O₃
  • Description:
    (2RS)-2-(4-Hydroxyphenyl)propionic acid

 

 

Biochemical and physiol actions are 2-(4-Hydroxyphenyl)propionic acid is a polyphenol metabolite detected in biological fluids. This compound belongs to the class of organic compounds known as phenylpropanoic acids. These are compounds with a structure containing a benzene ring conjugated to a propanoic acid.