Supelco® Organic Reference Standard, (+)-ʟ-Alliin
Katalog # 72805-10MG
Leverantör: Merck
CAS-nummer:
Specifikationer
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:556-27-4
- Description:(+)-ʟ-Alliin
- Storage conditions:2…8 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):(S)-S-Allyl-ʟ-cysteine sulfoxide; (S)-ʟ-3-(Allylsulfinyl)alanine
- Molecular weight:177,22
Specifikationer
Om denna produkt
- GHS signal: GHS07
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338