Order Entry
Sweden
ContactUsLinkComponent
Goggles, ultravision wide-vision gas-tight
Goggles, ultravision wide-vision gas-tight
Catalog # UVEX9301.613
Supplier:  UVEX
undefined
Goggles, ultravision wide-vision gas-tight
Catalog # UVEX9301.613
Supplier:  UVEX
Specifications
  • Type:
    Anti-UV, anti-scratch, anti-fog, gas-tight
  • Colour:
    Yellow
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
Classic, ergonomic wide-vision goggle with unrestricted side visibility.
  • Anti-fog coating on the inside, extremely resistant to scratches and chemicals on the outside
  • Gas-tight version without a ventilation system
  • Large goggle body, can also comfortably be worn as overspecs
  • Yellow frame
  • Clear PC lens
The uvex ultravision features a comfortable and ergonomic design and offers unrestricted side visibility. This flexible wide-vision goggle is also suitable for use as overspecs. The uvex supravision excellence coating provides anti-fog protection on the inside and makes the outside extremely resistant to chemicals and scratches. Lenses can be replaced.