Pesticide standard, Pyriftalid, reference material, Honeywell Fluka™
HONCR1681-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:135186-78-6
- Description:Pyriftalid
- Formula:C₁₅H₁₄N₂O₄S
- Storage temperature:Ambient
- Description:Pyriftalid
- Form:Solid
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.