Organic analytical standards, 2,3-Butanedione, reference material, Honeywell Fluka™
HONCR1089-2ML
:
- Pack type:Glass Bottle
- Pk:2 mL
- Standards:Analytical Standard
- CAS No.:431-03-8
- Description:2,3-Butanedione
- Melting point:–4 … –2 °C
- Formula:CH₃COCOCH₃
- Storage temperature:Ambient
- Description:2,3-Butanedione
- Flash point:7 °C
- Boiling point:88 °C
- Form:Neat
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.