Supelco® Organic Reference Standard, EPA 8270 Calibration Mix 1
Catalog # 5M00720
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 µg/ml each component in dichloromethane : benzene (3:1)
- Pk:1
- Standards:Analytical standard
- Description:EPA 8270 Calibration mix 1
- Storage conditions:–10...–25 °C
- Hazard class description:H225 - H302 + H332 - H304 - H315 - H319 - H336 - H340 - H350 - H372 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
Specifications
About this item
- GHS signal: GHS08, GHS07, GHS09
Caution: P210 - P273 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P312 - P331