Supelco® Organic Reference Standard, EPA 525 Fortification Solution A
Catalog # 48230-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 μg/ml each component in dichloromethane
- Pk:1
- Standards:Certified reference material
- Description:EPA 525 fortification solution A
- Storage conditions:2…8 °C
- Hazard class description:H315 - H319 - H336 - H350 - H411
- Shipping conditions:STD
- M-Clarity quality level:MQ100
Specifications
About this item
- GHS signal: GHS08, GHS09
Caution: P202 - P261 - P273 - P302 + P352 - P305 + P351 + P338 - P308 + P313