Order Entry
Sweden
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, EPA TCL Base/Neutrals Mix 2
  48120-U
 :  Merck
 :  
Supelco® Organic Reference Standard, EPA TCL Base/Neutrals Mix 2
  48120-U
 :  Merck
 :  

 

  • Concentration:
    2000 μg/ml each component in methylene chloride
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    EPA TCL Base/Neutrals mix 2
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H315 - H319 - H331 - H336 - H350 - H410
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100

 

 

  • GHS signal: GHS08, GHS07, GHS09
 : P202 - P273 - P302 + P352 - P304 + P340 + P311 - P305 + P351 + P338 - P308 + P313