Order Entry
Sweden
ContactUsLinkComponent
Supelco® Organic Reference Standard, EPA 8270 Matrix Spike Mix
Supelco® Organic Reference Standard, EPA 8270 Matrix Spike Mix
Catalog # 46854-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, EPA 8270 Matrix Spike Mix
Catalog # 46854-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    In methanol (varied conc.)
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    EPA 8270 Matrix spike mix
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H350 - H370 - H412
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Flash point:
    11 °C

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08, GHS09
Caution: P210 - P273 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311