Organophosphorus pesticides Mix A, certified reference material, Supelco®
Catalog # 48391.
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 mug/mL each component in hexane: acetone (9:1)
- Pk:1
- Description:Organophosphorus Pesticides Mix A
- Type:Certified reference material