Order Entry
Sweden
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Thiamethoxam
Supelco® PESTANAL® Organic Reference Standard, Thiamethoxam
Catalog # 37924-100MG-R
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Thiamethoxam
Catalog # 37924-100MG-R
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    153719-23-4
  • Description:
    Thiamethoxam
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H228 - H302 - H361fd - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD03792862
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3-(2-Chloro-5-thiazolylmethyl)tetrahydro-5-methyl-N-nitro-4H-1,3,5-oxadiazin-4-imine
  • Formula:
    C₈H₁₀ClN₅O₃S
  • Molecular weight:
    291,71
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07, GHS09
Caution: P202 - P210 - P240 - P273 - P301 + P312 - P308 + P313