Order Entry
Sweden
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Tebutam
Supelco® PESTANAL® Organic Reference Standard, Tebutam
Catalog # 36566-250MG
Leverantör:  Merck
CAS-nummer:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Tebutam
Catalog # 36566-250MG
Leverantör:  Merck
CAS-nummer:  

Specifikationer

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    35256-85-0
  • Description:
    Tebutam
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00145351
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    N-Benzyl-N-isopropylpivalamide
  • Formula:
    C₁₅H₂₃NO
  • Molecular weight:
    233,35
  • Flash point:
    80 °C - closed cup

Specifikationer

Om denna produkt