Supelco® PESTANAL® Organic Reference Standard, Propham
Catalog # 45641-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:122-42-9
- Description:Propham
- Storage conditions:2...30 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00026382
- M-Clarity quality level:MQ100
- Synonym(s):IPC; Isopropyl phenylcarbamate
- Formula:C₁₀H₁₃NO₂
- Molecular weight:179,22
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07
Caution: P301 + P312 + P330