Organic reference standard, Perylene-d12 solution, certified reference material, Supelco®
Catalog # 48081.
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 µg/ml in dichloromethane
- Pk:1
- Standards:Certified reference material
- CAS No.:1520-96-3
- Description:Perylene-d12 solution
- Formula:C₂₀H₁₂
- Storage temperature:2...8 °C
- Description:Perylene-d12 solution
- Pk Info:1 ml/ampule