Order Entry
Sweden
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, N-(6-Chloro-3-Pyridylmethyl)-N-Cyano-N-Methylacetamidine
  33674-100MG-R
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, N-(6-Chloro-3-Pyridylmethyl)-N-Cyano-N-Methylacetamidine
  33674-100MG-R
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    160430-64-8
  • Description:
    N-(6-Chloro-3-pyridylmethyl)-N-cyano-N-methylacetamidine
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H301 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD06201842
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Acetamiprid, sum of isomers
  • Melting point:
    100...102 °C
  • Formula:
     C₁₀H₁₁ClN₄
  • Molecular weight:
    222,67
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS06
 : P264 - P270 - P273 - P301 + P310 - P391 - P405