Organic reference standard, Methyl octacosanoate, analytical standard, Supelco®
Catalog # 74701-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:Analytical standard
- CAS No.:55682-92-3
- Description:Methyl octacosanoate
- M-Clarity quality level:MQ100
- Melting point:67...69 °C
- Formula:C₂₉H₅₈O₂
- Description:Methyl octacosanoate