Supelco® PESTANAL® Organic Reference Standard, Genite
45519-250MG
: Merck
:
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:97-16-5
- Description:Genite
- Storage conditions:2...30 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00055413
- M-Clarity quality level:MQ100
- Synonym(s):2,4-Dichlorophenyl benzenesulfonate
- Formula:C₁₂H₈Cl₂O₃S
- Molecular weight:303,16
- Flash point:Not applicable
- GHS signal: GHS07
: P301 + P312 + P330