Organic reference standard, Formaldehyde Oxazolidine solution, certified reference material, Supelco®
48414.
: Merck
:
- Concentration:2000 µg/ml in toluene
- Pk:1 Ampoul
- Standards:Certified reference material
- CAS No.:108-88-3
- Description:Formaldehyde Oxazolidine solution
- Storage temperature:2...8 °C
- Description:Formaldehyde Oxazolidine solution
- Pk Info:1 ml/ampule
: ISO 34:2009 and ISO/IEC 17025:2005.