Order Entry
Sweden
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Dinotefuran
  32499-50MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Dinotefuran
  32499-50MG
 :  Merck
 :  

 

  • Pk:
    50 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    165252-70-0
  • Description:
    Dinotefuran
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H319
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD06795001
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (RS)-N-Methyl-N′-nitro-N″-[(tetrahydro-3-furanyl)methyl]guanidine
  • Formula:
    C₇H₁₄N₄O₃
  • Molecular weight:
    202,21
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS07
 : P264 - P280 - P305 + P351 + P338 - P337 + P313