Supelco® PESTANAL® Organic Reference Standard, Azimsulfuron
Catalog # 32521-25MG
Supplier: Merck
CAS Number:
Specifications
- Förp.:25 mg
- Standard:PESTANAL®, Analytical standard
- CAS-nr:120162-55-2
- Beskrivning:Azimsulfuron
- Storage conditions:2...30 °C
- Faroklassbeskrivning:H410
- Shipping conditions:STD
- MDL No.:MFCD02102190
- M-Clarity quality level:MQ100
- Synonym(s):N-{[(4,6-Dimethoxy-2-pyrimidinyl)amino]carbonyl}-1-methyl-4-(2-methyl-H-tetrazol-5-yl)-1H-pyrazole-5-sulfonamide
- Formel:C₁₃H₁₆N₁₀O₅S
- Molekylvikt:424,40
- Flampunkt:Not applicable
Specifications
About this item
- GHS signal: GHS09
Caution: P273