Organic reference standard, Aroclor 1260 solution, certified reference material, Supelco®
Catalog # 48735.
Supplier: Merck
CAS Number:
Specifications
- Concentration:500 mg/kg in transformer oil
- Pk:1
- Standards:Certified reference material
- CAS No.:11096-82-5
- Description:Aroclor 1260 solution
- Storage temperature:RT
- Description:Aroclor 1260 solution
- Pk Info:5 ml/ampule