Order Entry
Sweden
ContactUsLinkComponent
 
Organic reference standard, Aroclor 1254 solution, certified reference material, Supelco®
 :  Merck
 
Organic reference standard, Aroclor 1254 solution, certified reference material, Supelco®
 :  Merck

 

  • CAS No.:
    11097-69-1
  • Storage temperature:
    RT
  • Description:
    Aroclor 1254 solution