Order Entry
Sweden
ContactUsLinkComponent
Organic reference standard, 4-Methyl-1H-benzotriazole, analytical standard, Supelco®
Organic reference standard, 4-Methyl-1H-benzotriazole, analytical standard, Supelco®
Catalog # 14593-50MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 4-Methyl-1H-benzotriazole, analytical standard, Supelco®
Catalog # 14593-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    29878-31-7
  • Description:
    4-Methyl-1H-benzotriazole
  • Melting point:
    141...146 °C
  • Formula:
    C₇H₇N₃
  • Description:
    4-Methyl-1H-benzotriazole

Specifications

About this item

4-Methyl-1H-benzotriazole is an additive in formulated aircraft deicing and anti-icing fluids (ADAFs) mixture. It was used as analytical standard for a study demonstrated to determine benzotriazoles and benzothiazoles in human urine samples using LC- ESI(+)MS/MS.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.