Supelco® Organic Reference Standard, (±)-γ-Hexabromocyclododecane
73709-25MG
: Merck
:
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:134237-52-8
- Description:(±)-γ-Hexabromocyclododecane
- Storage conditions:2...30 °C
- Hazard class description:H361 - H362 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):γ-HBCD; (1RS,2RS,5RS,6SR,9SR,10RS)-1,2,5,6,9,10-Hexabromocyclododecane
- Melting point:208 - 214 °C
- Molecular weight:641,70
- GHS signal: GHS08, GHS09
: P202 - P260 - P263 - P264 - P273 - P308 + P313