Order Entry
Sweden
ContactUsLinkComponent
Membranes, AE, Whatman™
Membranes, AE, Whatman™
  WHAT10549916
undefined
Membranes, AE, Whatman™
  WHAT10549916

 

  • Type:
    AE98
  • Size:
    25 mm × 50 m
  • Thickness:
    120 μm
  • Pk:
    1

 

 

These are unbacked nitrocellulose membranes for use in a lateral flow assay.

  • Unbacked, allowing either belt or air side of the membrane to be used
  • Enables assay suitability tests of both air and belt side of the membrane
  • Constructed of 100% nitrocellulose

These AE membranes offer a higher level of purity and performance compared to post-treated materials. The AE membranes have been used extensively since the development of the original lateral-flow tests.

Capillary flow rate: 160 - 210 s/4 cm