Toluidine blue O, powder, Certifiable Grade
Catalog # E847-25G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Black to dark green fine powder
- Dye Content:> = 50 %
- Loss on Drying:< = 8 %
- Metachromatic Stain:Pass
- Solubility (0.016g, 2L water):Pass
Specification Test Results
About this item
:
:305.83 g/mol
Storage Temperature:Ambient
CAS Number:92-31-9
Formula:C₁₅H₁₆ClN₃S
MDL Number:MFCD00011934