Pharmaceutical analytical standard, Cefacetrile, reference material, Honeywell Fluka™
Catalog # HONCR1434-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:10206-21-0
- Description:Cefacetrile
- Formula:C₁₃H₁₃N₃O₆S
- Storage temperature:Ambient
- Description:Cefacetrile
- Freezing point:174...177 °C
Specifications
About this item
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results