ESD safety shoes, Velcro® fastening, uvex 2 xenova® S1P Trainer, 9553
Leverantör: UVEX
Certifikat
Om denna produkt
Metal-free, extremely lightweight, flexible and multifunctional safety shoe made from breathable water resistant high-tech micro velour upper, soft padded tongue and collar, plus a dual-density PUR sole (oil and petrol resistant, anti-static) free of paint-wetting impairment substances, silicones and plasticizers.
- Climazone technology for improved comfort
- Double individually adjustable velcro
- Breathable distance mesh lining
- Removable anti-static comfort insole with moisture control system; with a moisture-wicking system
- Anti-slip sole with shock-absorption at heel and front of the foot
- Virtually seamless design eliminates potential pressure points and blisters
- 100% metal-free xenova® toecap
- Flexible, metal-free, puncture resistant midsole
- Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome, made from synthetic textiles.
Colour: Black/blue
Certifications: EN 61340 (ESD); EN ISO 20345:2011; S1P, SRC