0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Cholesterol margarate
Cholesterol margarate ≥95%
Cholesterol margarate ≥95%
Cholesterol margarate ≥95%
Leverantör:
Cayman Chemical
Certifikat
Expand image
Cholesterol margarate ≥95%
Leverantör:
Cayman Chemical
Certifikat
Om denna produkt
CAS-nummer:
24365-37-5
:
Formel:
C₄₄H₇₈O₂
MDL-nummer:
MFCD00056438
:
639.11 g/mol
Förvaringstemperatur:
Freezer
Specifikationstestresultat
Purity:
≥95%
λmax:
201 nm
Specifikationstestresultat
Relaterad information
Documentation
Certifikat
Sök efter LOT-nummer
Documentation